Flt3-Ligand Human Recombinant produced in Sf9 insect cells is glycosylated, polypeptide chain containing 155 amino acids and having migrating on SDS-PAGE at 19kDa. Flt3-Ligand is purified by proprietary chromatographic techniques. 1 * 10 µG
Catalog # BIRBORB80326-10
Supplier: Biorbyt