ContactUsLinkComponent
Flt3-Ligand Human Recombinant produced in HEK293 cells is glycosylated, polypeptide chain, migrates as diffuse band on SDS-PAGE due to heterogeneous glycosylation at 30kDa. Flt3-Ligand is purified by proprietary chromatographic techniques. 1 * 10 µG
Catalog # BIRBORB80324-10
Supplier:  Biorbyt
Flt3-Ligand Human Recombinant produced in HEK293 cells is glycosylated, polypeptide chain, migrates as diffuse band on SDS-PAGE due to heterogeneous glycosylation at 30kDa. Flt3-Ligand is purified by proprietary chromatographic techniques. 1 * 10 µG
Catalog # BIRBORB80324-10
Supplier:  Biorbyt