ContactUsLinkComponent
FGF 22 Human Recombinant produced in E.coli is a single, non-glycosylated polypeptide chain containing 149 amino acids and having a molecular mass of 17.3 kDa. The FGF-22 is purified by chromatographic techniques. 1 * 2 µG
Catalog # BIRBORB80322-2
Supplier:  Biorbyt
FGF 22 Human Recombinant produced in E.coli is a single, non-glycosylated polypeptide chain containing 149 amino acids and having a molecular mass of 17.3 kDa. The FGF-22 is purified by chromatographic techniques. 1 * 2 µG
Catalog # BIRBORB80322-2
Supplier:  Biorbyt