ContactUsLinkComponent
Sincalide also called CCK-8 has molecular formula of C49H62N10O16S3, H-Asp-Tyr(SO3H)-Met-Gly-Trp-Met-Asp-Phe-NH2 having Mw of 1143.2 9 Dalton. 1 * 1 mg
Catalog # BIRBORB80756-1
Supplier:  Biorbyt
Sincalide also called CCK-8 has molecular formula of C49H62N10O16S3, H-Asp-Tyr(SO3H)-Met-Gly-Trp-Met-Asp-Phe-NH2 having Mw of 1143.2 9 Dalton. 1 * 1 mg
Catalog # BIRBORB80756-1
Supplier:  Biorbyt