ContactUsLinkComponent
Calcitonin Acetate (Salmon) is synthetic polypeptide of 32 amino acids in the same linear sequencethat is found in clacitonin of sal mon origin. Calcitonin Acetate (Salmon) has an amino acid sequence of: Cys-Ser-Asn-Leu-Ser-Thr-Cys-Val-Leu-G 1 * 25 mg
Catalog # BIRBORB80754-25
Supplier:  Biorbyt
Calcitonin Acetate (Salmon) is synthetic polypeptide of 32 amino acids in the same linear sequencethat is found in clacitonin of sal mon origin. Calcitonin Acetate (Salmon) has an amino acid sequence of: Cys-Ser-Asn-Leu-Ser-Thr-Cys-Val-Leu-G 1 * 25 mg
Catalog # BIRBORB80754-25
Supplier:  Biorbyt