ContactUsLinkComponent
Fibroblast Growth Factor -21 Mouse Recombinant produced in E.coli is a single, non-glycosylated, polypeptide chain containing 192 am ino acids and having a molecular mass of 21.2 kDa. The amino acid sequence of the recombinant human FGF21 is 1 * 2 µG
Catalog # BIRBORB80320-2
Supplier:  Biorbyt
Fibroblast Growth Factor -21 Mouse Recombinant produced in E.coli is a single, non-glycosylated, polypeptide chain containing 192 am ino acids and having a molecular mass of 21.2 kDa. The amino acid sequence of the recombinant human FGF21 is 1 * 2 µG
Catalog # BIRBORB80320-2
Supplier:  Biorbyt