FGF 2 Human Recombinant (FGF-2) produced in E.coli is a single, non-glycosylated, polypeptide chain containing 147 amino acids and h aving a molecular mass of 16539 Dalton.The FGF2 is purified by proprietary chromatographic techniques. 1 * 50 µG
Catalog # BIRBORB80310-50
Supplier: Biorbyt