ContactUsLinkComponent
Fibroblast Growth Factor -21 Human Recombinant produced in E.coli is a single, non-glycosylated, polypeptide chain containing 202 am ino acids (29-209) and having a molecular mass of 21.6 kDa. The FGF-21 is fused to 20 amino acid His Tga at 1 * 10 µG
Catalog # BIRBORB80318-10
Supplier:  Biorbyt
Fibroblast Growth Factor -21 Human Recombinant produced in E.coli is a single, non-glycosylated, polypeptide chain containing 202 am ino acids (29-209) and having a molecular mass of 21.6 kDa. The FGF-21 is fused to 20 amino acid His Tga at 1 * 10 µG
Catalog # BIRBORB80318-10
Supplier:  Biorbyt