ContactUsLinkComponent
Fibroblast Growth Factor -19 Human Recombinant produced in E.coli is a single, non-glycosylated, polypeptide chain containing 206 am ino acids and having a molecular mass of 23 kDa. The amino acid sequence of the recombinant human FGF19 is 1 1 * 5 µG
Catalog # BIRBORB80317-5
Supplier:  Biorbyt
Fibroblast Growth Factor -19 Human Recombinant produced in E.coli is a single, non-glycosylated, polypeptide chain containing 206 am ino acids and having a molecular mass of 23 kDa. The amino acid sequence of the recombinant human FGF19 is 1 1 * 5 µG
Catalog # BIRBORB80317-5
Supplier:  Biorbyt