Fibroblast Growth Factor -19 Human Recombinant produced in E.coli is a single, non-glycosylated, polypeptide chain containing 206 am ino acids and having a molecular mass of 23 kDa. The amino acid sequence of the recombinant human FGF19 is 1 1 * 25 µG
Catalog # BIRBORB80317-25
Supplier: Biorbyt