ContactUsLinkComponent
Pramlintide Synthetic is a single, non-glycosylated polypeptide chain containing 37 amino acids, having a molecular mass of 3949.4 D alton and a molecular formula of C171H267N51O53S2. 1 * 25 mg
Catalog # BIRBORB80749-25
Supplier:  Biorbyt
Pramlintide Synthetic is a single, non-glycosylated polypeptide chain containing 37 amino acids, having a molecular mass of 3949.4 D alton and a molecular formula of C171H267N51O53S2. 1 * 25 mg
Catalog # BIRBORB80749-25
Supplier:  Biorbyt