ContactUsLinkComponent
FGF acidic Mouse Recombinant (FGF-1) produced in E.coli is a single, non-glycosylated, polypeptide chain containing 141 amino acids and having a molecular mass of 15.9 kDa.The FGF acidic is purified by proprietary chromatographic techniques 1 * 10 µG
Catalog # BIRBORB80307-10
Supplier:  Biorbyt
FGF acidic Mouse Recombinant (FGF-1) produced in E.coli is a single, non-glycosylated, polypeptide chain containing 141 amino acids and having a molecular mass of 15.9 kDa.The FGF acidic is purified by proprietary chromatographic techniques 1 * 10 µG
Catalog # BIRBORB80307-10
Supplier:  Biorbyt