CD105 Mouse Recombinant extracellular domain produced in baculovirus is homodimeric, glycosylated, Polypeptide containing 581 amino acids and having a molecular mass of 61 kDa but as result of glycosylation, migrates at 75-85 kDa under redu 1 * 100 µG
Catalog # BIRBORB80305-100
Supplier: Biorbyt