ContactUsLinkComponent
CD105 Human Recombinant extracellular domain produced in baculovirus is homodimeric, glycosylated, Polypeptide containing 586 amino acids and having a molecular mass of 61 kDa but as result of glycosylation, migrates at 90 kDa under reducin 1 * 2 µG
Catalog # BIRBORB80303-2
Supplier:  Biorbyt
CD105 Human Recombinant extracellular domain produced in baculovirus is homodimeric, glycosylated, Polypeptide containing 586 amino acids and having a molecular mass of 61 kDa but as result of glycosylation, migrates at 90 kDa under reducin 1 * 2 µG
Catalog # BIRBORB80303-2
Supplier:  Biorbyt