ContactUsLinkComponent
Insulin Human Recombinant produced in E.coli is two chain, non-glycosylated polypeptide chain containing 51 amino acids and having a molecular mass of 5807 Dalton. Insulin is purified by proprietary chromatographic techniques. 1 * 25 mg
Catalog # BIRBORB80481-25
Supplier:  Biorbyt
Insulin Human Recombinant produced in E.coli is two chain, non-glycosylated polypeptide chain containing 51 amino acids and having a molecular mass of 5807 Dalton. Insulin is purified by proprietary chromatographic techniques. 1 * 25 mg
Catalog # BIRBORB80481-25
Supplier:  Biorbyt