ContactUsLinkComponent
Human IL33 Recombinant produced in E.coli is a single, non-glycosylated, Polypeptide chain containing 159 amino acids C-terminal fra gment (112-270) having molecular weight of 20.5kDa and fused with 4.5kDa amino-terminal hexahistidine tag. T 1 * 10 µG
Catalog # BIRBORB80480-10
Supplier:  Biorbyt
Human IL33 Recombinant produced in E.coli is a single, non-glycosylated, Polypeptide chain containing 159 amino acids C-terminal fra gment (112-270) having molecular weight of 20.5kDa and fused with 4.5kDa amino-terminal hexahistidine tag. T 1 * 10 µG
Catalog # BIRBORB80480-10
Supplier:  Biorbyt