EBI3 Human Recombinant produced in E.coli is a single, non-glycosylated polypeptide chain containing 209 amino acids and having a mo lecular mass of 23.3 kDa. The Human EBI3 is purified by proprietary chromatographic techniques. 1 * 2 µG
Catalog # BIRBORB80295-2
Supplier: Biorbyt