ContactUsLinkComponent
Betacellulin Bovine Recombinant produced in E.coli is a single, non-glycosylated, polypeptide chain containing 80 amino acids and ha ving a molecular mass of 9003 Dalton. Betacellulin Bovine Recombinant is purified by proprietary chromatogra 1 * 20 µG
Catalog # BIRBORB80286-20
Supplier:  Biorbyt
Betacellulin Bovine Recombinant produced in E.coli is a single, non-glycosylated, polypeptide chain containing 80 amino acids and ha ving a molecular mass of 9003 Dalton. Betacellulin Bovine Recombinant is purified by proprietary chromatogra 1 * 20 µG
Catalog # BIRBORB80286-20
Supplier:  Biorbyt