ContactUsLinkComponent
BNP Peptide Recombinant Human produced in E.coli is a single, non-glycosylated, polypeptide chain containing 129 amino acids and hav ing a molecular mass of 14 kDa. NPPB is purified by proprietary chromatographic techniques. 1 * 2 µG
Catalog # BIRBORB80284-2
Supplier:  Biorbyt
BNP Peptide Recombinant Human produced in E.coli is a single, non-glycosylated, polypeptide chain containing 129 amino acids and hav ing a molecular mass of 14 kDa. NPPB is purified by proprietary chromatographic techniques. 1 * 2 µG
Catalog # BIRBORB80284-2
Supplier:  Biorbyt