C-kit ligand, the recently identified ligand for the kit tyrosine kinase receptor, is mapped to the mouse S1 locus. This pleiotropic cytokine, alternately known as stem cell factor (SCF), mast cell growth factor (MGF) and steel-factor (SLF), 1 * 2 µG
Catalog # BIRBORB107800-2
Supplier: Biorbyt