ContactUsLinkComponent
Interleukin-36 (IL-36) is a pro-inflammatory cytokine which plays an important role in the pathophysiology of several diseases.IL-36 D, IL-36G, and IL-36H (formerly IL-1F6, IL-1F8, and IL-1F9) are IL-1 family members that signal through the 1 * 10 µG
Catalog # BIRBORB107764-10
Supplier:  Biorbyt
Interleukin-36 (IL-36) is a pro-inflammatory cytokine which plays an important role in the pathophysiology of several diseases.IL-36 D, IL-36G, and IL-36H (formerly IL-1F6, IL-1F8, and IL-1F9) are IL-1 family members that signal through the 1 * 10 µG
Catalog # BIRBORB107764-10
Supplier:  Biorbyt