ContactUsLinkComponent
Interleukin-8 (C-X-C motif chemokine 8) is coded by the interleukin 8 gene on the chromosome 5 in Rhesus macaque (Macacamulatta), wh ich is not only 94% identical to human IL-8 but also structurally and functionally similar with the human eq 1 * 25 µG
Catalog # BIRBORB107752-25
Supplier:  Biorbyt
Interleukin-8 (C-X-C motif chemokine 8) is coded by the interleukin 8 gene on the chromosome 5 in Rhesus macaque (Macacamulatta), wh ich is not only 94% identical to human IL-8 but also structurally and functionally similar with the human eq 1 * 25 µG
Catalog # BIRBORB107752-25
Supplier:  Biorbyt