Interferon-Omega (IFN-Omega) coded by IFNW1 gene in human, is a number of the type I interferon family, which includes IFN-a,IFN-bet a, and IFN-Omega. The IFNAR-1/IFNAR-2 receptor complex can help with the signal transduction, followed the a 1 * 20 G
Catalog # BIRBORB107738-20
Supplier: Biorbyt