ContactUsLinkComponent
Skimmer cone, copper, for 75cs. 1/pk. for special applications such as ultra trace analysis by cool mode and pt analysis by hot mode in hydrogen peroxide. use with ni sampling cone 1 * 1 items
Catalog # HEWLG3270-60643
Supplier:  Agilent
Skimmer cone, copper, for 75cs. 1/pk. for special applications such as ultra trace analysis by cool mode and pt analysis by hot mode in hydrogen peroxide. use with ni sampling cone 1 * 1 items
Catalog # HEWLG3270-60643
Supplier:  Agilent
Frequently Bought Together