ContactUsLinkComponent
Low flow concentric glass nebulizer (micromist). 1/pk. provides aspirated free flow of .2 ml/min. includes sample and gas inlet fitt ings 1 * 1 items
Catalog # VARI190064300
Supplier:  VARIAN
Low flow concentric glass nebulizer (micromist). 1/pk. provides aspirated free flow of .2 ml/min. includes sample and gas inlet fitt ings 1 * 1 items
Catalog # VARI190064300
Supplier:  VARIAN
Frequently Bought Together