ContactUsLinkComponent
Organics sis kit, radial icp w/inert s/c. for use with organic solvents. incl v-groove neb, inert sturman-masters s/chamber, torch ( 1.4 mm id injector) + all necessary marprene/solvent tubing 1 * 1 items
Catalog # VARI9910126600
Supplier:  VARIAN
Organics sis kit, radial icp w/inert s/c. for use with organic solvents. incl v-groove neb, inert sturman-masters s/chamber, torch ( 1.4 mm id injector) + all necessary marprene/solvent tubing 1 * 1 items
Catalog # VARI9910126600
Supplier:  VARIAN
Frequently Bought Together