ContactUsLinkComponent
Aromatics sis kit radial icp w/inert s/c. for use with aromatic solvents. incl v-groove neb., inert sturman-masters s/chamber, torch (1.4 mm id injector) + all necessary viton/solvent tubing 1 * 1 items
Catalog # VARI9910127000
Supplier:  VARIAN
Aromatics sis kit radial icp w/inert s/c. for use with aromatic solvents. incl v-groove neb., inert sturman-masters s/chamber, torch (1.4 mm id injector) + all necessary viton/solvent tubing 1 * 1 items
Catalog # VARI9910127000
Supplier:  VARIAN
Frequently Bought Together