ContactUsLinkComponent
EC HPLC column EC 80/4 NUCLEOSIL 120-3 C18 Length: 80 mm, ID: 4 mm pack of 1 1 * 1 items
Catalog # MANA720318.40
Supplier:  MACHEREY-NAGEL
EC HPLC column EC 80/4 NUCLEOSIL 120-3 C18 Length: 80 mm, ID: 4 mm pack of 1 1 * 1 items
Catalog # MANA720318.40
Supplier:  MACHEREY-NAGEL
Frequently Bought Together