ContactUsLinkComponent
Paper Bag / PP PET 800x900mm Peelable pouch With 2 sterilization indicators : steam and EO 1 face paper (MDP15) 1 face blue PET-PP (MDL1) 800 x 900mm Box of 100 PCs. 1 * 1 Each
Catalog # BSNC121013
Supplier:  Basan
Paper Bag / PP PET 800x900mm Peelable pouch With 2 sterilization indicators : steam and EO 1 face paper (MDP15) 1 face blue PET-PP (MDL1) 800 x 900mm Box of 100 PCs. 1 * 1 Each
Catalog # BSNC121013
Supplier:  Basan
Frequently Bought Together