ContactUsLinkComponent
Mop system with tank Trapezium mop head with an Tank for dosage detergent or disinfectant Empty Weight 1.2 Kg Tank capacity 1 liter 1 * 1 Each
Catalog # BSNC323229
Supplier:  Basan
Mop system with tank Trapezium mop head with an Tank for dosage detergent or disinfectant Empty Weight 1.2 Kg Tank capacity 1 liter 1 * 1 Each
Catalog # BSNC323229
Supplier:  Basan
Frequently Bought Together