ContactUsLinkComponent
Nitrile GL 28cm Dermatril 742LR blue *** cleanroom class 100*** (bag = double packed) Colour blue, powderfree, length 280 mm, thickness 0,11 mm, 1 box = 1 unit = 50 pieces 10 box = 1 carton length 280 mm, thickness 0,11 mm, *** cleanroom class 100*** 1 * 500 items
Catalog # KCHL074209082C
Supplier:  KCL by Honeywell
Nitrile GL 28cm Dermatril 742LR blue *** cleanroom class 100*** (bag = double packed) Colour blue, powderfree, length 280 mm, thickness 0,11 mm, 1 box = 1 unit = 50 pieces 10 box = 1 carton length 280 mm, thickness 0,11 mm, *** cleanroom class 100*** 1 * 500 items
Catalog # KCHL074209082C
Supplier:  KCL by Honeywell
Frequently Bought Together