ContactUsLinkComponent
Flask, capacity 100ml, round bottom, diameter 64mm, height 115mm, SJ 29/32. 1 * 10 items
Catalog # KAVA632426219102
Supplier:  Kavalier
Flask, capacity 100ml, round bottom, diameter 64mm, height 115mm, SJ 29/32. 1 * 10 items
Catalog # KAVA632426219102
Supplier:  Kavalier
Frequently Bought Together