PH4 BUFFER SOLUTION 1 * 500 mL
SENK788-11
: SENTEK
Frequently Bought Together
4'-Hydroxydiclofenac, Supelco®
: Merck
Ethanolamine, Supelco®
: Merck
Buffer solutions in sachets
: Hanna
Buffer Solutions
: Thermo Fisher Scientific
MULTI ELEMENT STD 75 BLANK OIL S-21:30 1 * 200 g
: LGC Standards PROMOCHEM
Pirimicarb Analytical standard, reference material, Fluka™
: Honeywell Chemicals
Conductivity standards, KCL
: Hach