Sucrose stearate 1 * 25 g
J66845.14
Frequently Bought Together
Water, HiPerSolv CHROMANORM® for HPLC
: VWR Chemicals
Acetone ≥99.5%, GPR RECTAPUR®
: VWR Chemicals
Ethanol 96% vol Ph. Eur.
: VWR Chemicals
Ethyl acetate ≥99.8%, HiPerSolv CHROMANORM® for HPLC
: VWR Chemicals
Ethanol, GPR RECTAPUR®
: VWR Chemicals
Blocking buffers
: Rockland Immunochemicals
SODIUM STEAROYL LACTYLATE 1 * 500 g
: Apollo Scientific