ContactUsLinkComponent
Human CENPP partial ORF ( NP_001012267, 1 A.A. - 98 A.A.) Recombinant protein with GST-TAG AT N-terminal. 1 * 25 µG
Catalog # ABNOH00401541-Q012
Supplier:  Abnova
Human CENPP partial ORF ( NP_001012267, 1 A.A. - 98 A.A.) Recombinant protein with GST-TAG AT N-terminal. 1 * 25 µG
Catalog # ABNOH00401541-Q012
Supplier:  Abnova
Frequently Bought Together