ContactUsLinkComponent
HLA-DPB1 (Human) Recombinant protein (P02). Human HLA-DPB1 full-length ORF ( AAH07963, 30 A.A. - 258 A.A.) Recombinant protein with GST-TAG AT N-terminal. 1 * 25 µG
Catalog # ABNOH00003115-P022
Supplier:  Abnova
HLA-DPB1 (Human) Recombinant protein (P02). Human HLA-DPB1 full-length ORF ( AAH07963, 30 A.A. - 258 A.A.) Recombinant protein with GST-TAG AT N-terminal. 1 * 25 µG
Catalog # ABNOH00003115-P022
Supplier:  Abnova
Frequently Bought Together