ContactUsLinkComponent
Laboratory Chair Neon, Easy To Clean, Washable, Upholstery Made Of Pu-Foam, Grey, Backrest Height Adjustment, Seat Height Adjustment With A Gasspring From 450-620 Mm, 5-Star Base Made Of Aluminium, Powder Coated Black, With Stop-And-Go-Castors, With 1 * 1 items
Catalog # BIMO57120023280807
Supplier:  bimos
Laboratory Chair Neon, Easy To Clean, Washable, Upholstery Made Of Pu-Foam, Grey, Backrest Height Adjustment, Seat Height Adjustment With A Gasspring From 450-620 Mm, 5-Star Base Made Of Aluminium, Powder Coated Black, With Stop-And-Go-Castors, With 1 * 1 items
Catalog # BIMO57120023280807
Supplier:  bimos
Frequently Bought Together