Bismuth Indium Tin ingot (Field's metal) 1 * 100 g
Catalog # 46895.22
Supplier: Thermo Fisher Scientific
Bismuth Indium Tin ingot (Field's metal) 1 * 100 g
Catalog # 46895.22
Supplier: Thermo Fisher Scientific
Frequently Bought Together
Dichloromethane, dehydrated (max. 0.01% H₂O) ≥99% stabilised, GPR RECTAPUR® for synthesis
Supplier: VWR Chemicals
Ethanol 90% 90% (v/v), TechniSolv®, pure
Supplier: VWR Chemicals
Dichloromethane ≥99% stabilised, GPR RECTAPUR®
Supplier: VWR Chemicals
Tetrahydrofuran, extra dry 99.85% stabilised, AcroSeal®
Supplier: Thermo Fisher Scientific
1-Butylboronic acid, Supelco®
Supplier: Merck
Chlorobenzene 99.9% for HPLC
Supplier: Thermo Fisher Scientific
Methanol ≥99.8% for LC-MS
Supplier: Thermo Fisher Scientific
Dimethyl sulphoxide, extra dry over Molecular Sieve 99.7+%, AcroSeal®
Supplier: Thermo Fisher Scientific
Methanol ≥99.8% ACS, meets analytical specifications of Reagent Ph. Eur.
Supplier: Thermo Fisher Scientific
Isobutanol ≥99%
Supplier: Thermo Fisher Scientific
n-Pentane, anhydrous 99+%, extra pure
Supplier: Thermo Fisher Scientific
Ethanol (taxed), pure for molecular biology (200 Proof), Riedel-de Haën™
Supplier: Honeywell Chemicals
Ethanol absolute, BAKER ANALYZED® ACS, J.T.Baker®
Supplier: Avantor
Acetone ≥99.5%, Laboratory Reagent
Supplier: Honeywell Chemicals
Chloroform ≥99.5% stabilised
Supplier: Honeywell Chemicals