ContactUsLinkComponent
recombinant/native protein, ptgr1 (human) recombinant protein, human ptgr1 (np_036344, 1 a.a. - 329 a.a. ) full-length recombinant p rotein with his tag expressed in escherichia coli. 1 * 100 µG
Catalog # ABNOP5854
Supplier:  Abnova
recombinant/native protein, ptgr1 (human) recombinant protein, human ptgr1 (np_036344, 1 a.a. - 329 a.a. ) full-length recombinant p rotein with his tag expressed in escherichia coli. 1 * 100 µG
Catalog # ABNOP5854
Supplier:  Abnova