ContactUsLinkComponent
recombinant/native protein, crygc (human) recombinant protein, human crygc (np_066269, 1 a.a. - 174 a.a.) full-length recombinant pr otein with his tag expressed in escherichia coli. 1 * 100 µG
Catalog # ABNOP5395
Supplier:  Abnova
recombinant/native protein, crygc (human) recombinant protein, human crygc (np_066269, 1 a.a. - 174 a.a.) full-length recombinant pr otein with his tag expressed in escherichia coli. 1 * 100 µG
Catalog # ABNOP5395
Supplier:  Abnova