ContactUsLinkComponent
recombinant/native protein, fabp9 (human) recombinant protein, human fabp9 (np_001073995, 1 a.a. - 132 a.a.) full-length recombinant protein with his tag expressed in escherichia coli. 1 * 100 µG
Catalog # ABNOP5394
Supplier:  Abnova
recombinant/native protein, fabp9 (human) recombinant protein, human fabp9 (np_001073995, 1 a.a. - 132 a.a.) full-length recombinant protein with his tag expressed in escherichia coli. 1 * 100 µG
Catalog # ABNOP5394
Supplier:  Abnova