recombinant protein, tnfrsf10a (human) recombinant protein, purified tnfrsf10a (aah12866.1 103 a.a. - 239 a.a.) human recombinant pr otein with his-flag-strepii tag at n-terminus expressed in human cells. 1 * 25 µG
Catalog # ABNOH00008797-H01
Supplier: Abnova