ContactUsLinkComponent
recombinant protein, tnfrsf11a (human) recombinant protein, purified tnfrsf11a (np_003830.1 30 a.a. - 212 a.a.) human recombinant pr otein with his-flag-strepii tag at n-terminus expressed in human cells. 1 * 25 µG
Catalog # ABNOH00008792-H01
Supplier:  Abnova
recombinant protein, tnfrsf11a (human) recombinant protein, purified tnfrsf11a (np_003830.1 30 a.a. - 212 a.a.) human recombinant pr otein with his-flag-strepii tag at n-terminus expressed in human cells. 1 * 25 µG
Catalog # ABNOH00008792-H01
Supplier:  Abnova