ContactUsLinkComponent
recombinant protein, edar (human) recombinant protein, purified edar (aah93872.1, 27 a.a. - 183 a.a.) human recombinant protein with his-flag-strepii tag at n-terminus expressed in human cells. 1 * 25 µG
Catalog # ABNOH00010913-H01
Supplier:  Abnova
recombinant protein, edar (human) recombinant protein, purified edar (aah93872.1, 27 a.a. - 183 a.a.) human recombinant protein with his-flag-strepii tag at n-terminus expressed in human cells. 1 * 25 µG
Catalog # ABNOH00010913-H01
Supplier:  Abnova