ContactUsLinkComponent
recombinant protein, srpk2 (human) recombinant protein (p04), human srpk2 full-length orf (aah35214.1, 1 a.a. - 688 a.a.) recombinan t protein with gst-psts1 tag at n-terminal. 1 * 10 µG
Catalog # ABNOH00006733-P04
Supplier:  Abnova
recombinant protein, srpk2 (human) recombinant protein (p04), human srpk2 full-length orf (aah35214.1, 1 a.a. - 688 a.a.) recombinan t protein with gst-psts1 tag at n-terminal. 1 * 10 µG
Catalog # ABNOH00006733-P04
Supplier:  Abnova