ContactUsLinkComponent
recombinant protein, cd80 (human) recombinant protein, purified cd80 (np_005182.1, 35 a.a. - 242 a.a.) human recombinant protein wit h his-flag-strepii tag at n-terminus expressed in human cells. 1 * 25 µG
Catalog # ABNOH00000941-H01
Supplier:  Abnova
recombinant protein, cd80 (human) recombinant protein, purified cd80 (np_005182.1, 35 a.a. - 242 a.a.) human recombinant protein wit h his-flag-strepii tag at n-terminus expressed in human cells. 1 * 25 µG
Catalog # ABNOH00000941-H01
Supplier:  Abnova