Etoposide
100 mg
Supplier: Thermo Fisher Scientific
Jump to:
Product Details & Documents
CAS No.: 33419-42-0 Molecular Formula: C29H32O13 Formula Weight: 588.56 Physical Form: Powder Appearance: White Melting Point: 236-251° Optical Rotation: -107° (c=1 in methanol/dichloromethane 1:9) MDL No.: MFCD00869325
Applications: Topoisomerase inhibitor and inducer of apoptosis
Synonyms: (5R,5aR,8aR,9S)-9-(((4aR,6R,7R,8R,8aS)-7,8-Dihydroxy-2-methylhexahydropyrano[3,2-d][1,3]dioxin-6-yl)oxy)-5-(4-hydroxy-3,5-dimethoxyphenyl)-5,5a,8a,9-tetrahydrofuro[3',4':6,7]naphtho[2,3-d][1,3]dioxol-6(8H)-one, 4'-Demethylepipodophyllotoxin 9-(4,6-O-Ethylidene-beta-D-glucopyranoside)
- CAS Number:33419-42-0
- Formula:C₂₉H₃₂O₁₃
- MDL Number:MFCD00869325
- Molecular Weight:588.56 g/mol
- Merck Index:14,3886
- Melting Pt:236…251 °C
- Storage Temperature:Ambient
- EINECS:251-509-1
Specifications
Specifications
- Pk:100 mg



