Thiomersal
Glass bottle for solids, 10 g
Supplier: Thermo Fisher Scientific
Jump to:
Product Details & Documents
Synonyms: Ethylmercurithiosalicylic acid sodium salt, Sodium ethylmercurithiosalicylate, Thimerosal, Merthiolat, 2-[(Ethylmercury)thio]benzoic acid sodium salt, 2-(Ethylmercuriomercapto)benzoic acid sodium salt, 2-(Ethylmercuriothio)benzoic acid sodium salt
- CAS Number:54-64-8
- Formula:2-(C₂H₅HgS)C₆H₄CO₂Na
- MDL Number:MFCD00013062
- Molecular Weight:404.82 g/mol
- Merck Index:14,9317
- Melting Pt:234…237 °C
- Flash Pt:250 °C (482 °F)
- Storage Temperature:Ambient
- EINECS:200-210-4
- UN:2025
- ADR:6.1,II
Documents
Specifications
Specifications
- Pack type:Glass bottle for solids
- Pk:10 g
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



