ContactUsLinkComponent
FTA-1 ASPIRATOR WITH INTEGRAL PUMP AND TRAP FLASK 1L, -500MBAR VACUUM, 8 CHANNEL OPTION, HYDROPHOBIC FILTER 1 * 1 items
Catalog # GRANFTA-1EURO
Supplier:  GRANT INSTRUMENTS
undefined
FTA-1 ASPIRATOR WITH INTEGRAL PUMP AND TRAP FLASK 1L, -500MBAR VACUUM, 8 CHANNEL OPTION, HYDROPHOBIC FILTER 1 * 1 items
Catalog # GRANFTA-1EURO
Supplier:  GRANT INSTRUMENTS