ContactUsLinkComponent
11 COMPOSES 2000 MG/L CHAQUE Benzene [CAS:71-43-2] Ethylbenzene [CAS:100-41-4] Toluene [CAS:108-88-3] o-Xylene [CAS:95-47-6] m-Xylene [CAS:108-38-3] p-Xylene [CAS:106-42-3] Chlorobenzene [CAS:108-90-7] 1,2-Dichlorobenzene [CAS:95-50-1] 1 * 10 Ampoul
Catalog # 89263.001
Supplier:  VWR Collection
11 COMPOSES 2000 MG/L CHAQUE Benzene [CAS:71-43-2] Ethylbenzene [CAS:100-41-4] Toluene [CAS:108-88-3] o-Xylene [CAS:95-47-6] m-Xylene [CAS:108-38-3] p-Xylene [CAS:106-42-3] Chlorobenzene [CAS:108-90-7] 1,2-Dichlorobenzene [CAS:95-50-1] 1 * 10 Ampoul
Catalog # 89263.001
Supplier:  VWR Collection
Frequently Bought Together